Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107011.1 | NP_001100481.1 | 5 - 6 | 547 | 75 | |
GenBank mRNA | BC087071.1 | - | 5 - 6 | 724 | 75 | |
FQ213141.1 | - | 5 - 6 | 523 | 75 | ||
FQ220130.1 | - | 5 - 6 | 511 | 75 | ||
FQ229625.1 | - | 3 - 4 | 316 | 75 | ||
FQ229903.1 | - | 3 - 4 | 316 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |