Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053811.3 | NP_446263.2 | 3 - 4 | 673 | 87 | |
XM_006245893.2 | XP_006245955.1 | 2 - 3 | 1046 | 87 | ||
GenBank mRNA | AF294257.1 | - | 3 - 4 | 760 | 87 | |
CK840521.1 | - | 3 - 4 | 460 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |