Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053513.1 | NP_445965.1 | 12 - 13 | 1650 | 53 | |
XM_008768127.1 | XP_008766349.1 | 11 - 12 | 3055 | 53 | ||
XM_008768128.1 | XP_008766350.1 | 12 - 13 | 2724 | 53 | ||
XM_008768129.1 | XP_008766351.1 | 12 - 13 | 1736 | 53 | ||
XM_008768130.1 | XP_008766352.1 | 11 - 12 | 1581 | 53 | ||
GenBank mRNA | FQ211962.1 | - | 12 - 13 | 1835 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |