Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033683.1 | NP_001028855.1 | 4 - 5 | 910 | 76 | |
GenBank mRNA | BC101882.1 | - | 4 - 5 | 910 | 7929680 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |