Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105833.1 | NP_001099303.1 | 2 - 3 | 325 | 58 | |
GenBank mRNA | BC169002.1 | - | 2 - 3 | 268 | 7455281 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |