Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134729.1 | NP_001128201.1 | 3 - 4 | 182 | 147 | |
GenBank mRNA | BC168191.1 | - | 3 - 4 | 182 | 7107126 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |