Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144991.1 | NP_001138463.1 | 7 - 8 | 1264 | 72 | |
XM_006246562.2 | XP_006246624.2 | 7 - 8 | 1261 | 72 | ||
XM_006246563.2 | XP_006246625.2 | 7 - 8 | 1261 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |