Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247233.2 | XP_006247295.1 | 1 - 2 | 230 | 150 | |
XM_006247235.2 | XP_006247297.1 | 1 - 2 | 229 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |