Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760603.1 | XP_008758825.1 | 10 - 11 | 2492 | 102 | |
XM_008760604.1 | XP_008758826.1 | 10 - 11 | 1778 | 102 | ||
GenBank mRNA | BC111075.1 | - | 1 - 2 | 104 | 102 | |
FQ230057.1 | - | 3 - 4 | 486 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |