Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107648.1 | NP_001101118.1 | 2 - 3 | 221 | 139 | |
XM_006231847.2 | XP_006231909.1 | 2 - 3 | 253 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |