Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031097.1 | NP_112359.1 | 2 - 3 | 593 | 107 | |
GenBank mRNA | AY724503.1 | - | 2 - 3 | 765 | 107 | |
BC061718.1 | - | 2 - 3 | 623 | 5598331 | 107 | |
D87515.1 | - | 2 - 3 | 587 | 107 | ||
U61696.1 | - | 2 - 3 | 593 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |