Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053433.2 | NP_445885.2 | 4 - 5 | 709 | 90 | |
GenBank mRNA | AF286595.1 | - | 4 - 5 | 553 | 90 | |
BC087008.1 | - | 4 - 5 | 709 | 7129948 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |