Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250153.1 | XP_006250215.1 | 2 - 3 | 763 | 76 | |
XM_006250154.1 | XP_006250216.1 | 2 - 3 | 763 | 76 | ||
XM_008769624.1 | XP_008767846.1 | 2 - 3 | 763 | 76 | ||
GenBank mRNA | BC167054.1 | - | 2 - 3 | 521 | 7113939 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |