Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024902.1 | NP_001020073.1 | 8 - 9 | 974 | 107 | |
XM_006230564.2 | XP_006230626.1 | 8 - 9 | 1460 | 107 | ||
XM_006230565.2 | XP_006230627.1 | 8 - 9 | 1570 | 107 | ||
XM_006230566.2 | XP_006230628.1 | 8 - 9 | 1743 | 107 | ||
XM_006230569.2 | XP_006230631.1 | 8 - 9 | 1497 | 107 | ||
XM_006230570.2 | XP_006230632.1 | 6 - 7 | 707 | 107 | ||
XM_008760005.1 | XP_008758227.1 | 8 - 9 | 1742 | 107 | ||
GenBank mRNA | BC097444.1 | - | 8 - 9 | 974 | 7456428 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |