Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ighmbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031586.1 | NP_113774.1 | 12 - 13 | 1753 | 111 | |
GenBank mRNA | AF199411.1 | - | 12 - 13 | 1753 | 111 | |
BC099790.1 | - | 12 - 13 | 1949 | 7934952 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |