Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191622.1 | NP_001178551.1 | 11 - 12 | 1440 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |