Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106228.1 | NP_001099698.1 | 7 - 8 | 826 | 72 | |
XM_006228395.2 | XP_006228457.1 | 7 - 8 | 844 | 72 | ||
XM_006228396.1 | XP_006228458.1 | 7 - 8 | 804 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |