Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229179.2 | XP_006229241.1 | 2 - 3 | 348 | 76 | |
XM_006229180.2 | XP_006229242.1 | 2 - 3 | 348 | 76 | ||
XM_006229182.2 | XP_006229244.1 | 2 - 3 | 348 | 76 | ||
XM_008759449.1 | XP_008757671.1 | 2 - 3 | 348 | 76 | ||
XM_341847.7 | XP_341848.3 | 2 - 3 | 348 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |