Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004101.1 | NP_001004101.1 | 2 - 3 | 231 | 81 | |
XM_008759403.1 | XP_008757625.1 | 3 - 4 | 273 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |