Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127523.2 | NP_001120995.2 | 1 - 2 | 325 | 152 | |
GenBank mRNA | BC167102.1 | - | 1 - 2 | 325 | 8373935 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |