Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130821.1 | NP_570834.1 | 5 - 6 | 559 | 104 | |
GenBank mRNA | AB067679.1 | - | 5 - 6 | 559 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |