Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106719.2 | NP_001100189.1 | 4 - 5 | 424 | 68 | |
XM_006239979.2 | XP_006240041.1 | 3 - 4 | 340 | 68 | ||
XM_006239980.2 | XP_006240042.1 | 4 - 5 | 369 | 68 | ||
XM_006239981.2 | XP_006240043.1 | 3 - 4 | 551 | 68 | ||
GenBank mRNA | BC169104.1 | - | 4 - 5 | 424 | 7386477 | 68 |
FQ215419.1 | - | 3 - 4 | 259 | 68 | ||
FQ218542.1 | - | 3 - 4 | 301 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |