Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sntg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106720.1 | NP_001100190.1 | 4 - 5 | 471 | 56 | |
XM_006239967.2 | XP_006240029.1 | 8 - 9 | 752 | 56 | ||
XM_006239968.2 | XP_006240030.1 | 8 - 9 | 752 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |