Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239913.2 | XP_006239975.1 | 3 - 4 | 212 | 119 | |
GenBank mRNA | AY211264.1 | - | 3 - 4 | 151 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |