Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031573.1 | NP_113761.1 | 7 - 8 | 709 | 118 | |
GenBank mRNA | X07320.1 | - | 7 - 8 | 709 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |