Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127551.1 | NP_001121023.1 | 8 - 9 | 1075 | 121 | |
XM_006249175.2 | XP_006249237.1 | 8 - 9 | 1103 | 121 | ||
GenBank mRNA | BC166781.1 | - | 8 - 9 | 1075 | 7934984 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |