Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105925.1 | NP_001099395.1 | 7 - 8 | 1180 | 86 | |
XM_008769195.1 | XP_008767417.1 | 7 - 8 | 2463 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |