Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006957.2 | NP_001006958.1 | 3 - 4 | 347 | 82 | |
XM_006249134.1 | XP_006249196.1 | 3 - 4 | 570 | 82 | ||
GenBank mRNA | BC083818.1 | - | 3 - 4 | 339 | 7110979 | 82 |
CB718613.1 | - | 3 - 4 | 347 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |