Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144758.1 | NP_653359.1 | 6 - 7 | 1429 | 66 | |
GenBank mRNA | AB000280.1 | - | 6 - 7 | 1429 | 66 | |
BC087709.1 | - | 6 - 7 | 1428 | 7315369 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |