Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128193.2 | NP_001121665.1 | 3 - 4 | 434 | 139 | |
GenBank mRNA | BC166560.1 | - | 3 - 4 | 434 | 7314601 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |