Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106590.1 | NP_001100060.1 | 4 - 5 | 566 | 91 | |
XM_008762768.1 | XP_008760990.1 | 3 - 4 | 465 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Rattus norvegicus Probe spans exons |