Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106590.1 | NP_001100060.1 | 6 - 7 | 700 | 66 | |
XM_008762768.1 | XP_008760990.1 | 5 - 6 | 599 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |