Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106586.1 | NP_001100056.1 | 3 - 4 | 474 | 90 | |
XM_006236180.2 | XP_006236242.1 | 2 - 3 | 1615 | 90 | ||
XM_006236183.2 | XP_006236245.1 | 3 - 4 | 758 | 90 | ||
XM_006236184.2 | XP_006236246.1 | 3 - 4 | 498 | 90 | ||
XM_006236185.2 | XP_006236247.1 | 3 - 4 | 1549 | 90 | ||
XM_008762765.1 | XP_008760987.1 | 3 - 4 | 415 | 90 | ||
GenBank mRNA | FQ227572.1 | - | 3 - 4 | 454 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |