Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127298.2 | NP_001120770.2 | 12 - 13 | 1779 | 65 | |
XM_008762775.1 | XP_008760997.1 | 12 - 13 | 1881 | 65 | ||
XM_008762776.1 | XP_008760998.1 | 12 - 13 | 2180 | 65 | ||
GenBank mRNA | BC166785.1 | - | 12 - 13 | 1779 | 7935588 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |