Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127298.2 | NP_001120770.2 | 2 - 3 | 319 | 110 | |
XM_008762775.1 | XP_008760997.1 | 2 - 3 | 421 | 110 | ||
XM_008762776.1 | XP_008760998.1 | 2 - 3 | 720 | 110 | ||
GenBank mRNA | BC166785.1 | - | 2 - 3 | 319 | 7935588 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |