Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017456.1 | NP_001017456.1 | 4 - 5 | 757 | 92 | |
XM_006236201.2 | XP_006236263.1 | 5 - 6 | 913 | 92 | ||
GenBank mRNA | BC085351.1 | - | 4 - 5 | 757 | 7115168 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |