Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009647.1 | NP_001009647.1 | 3 - 4 | 353 | 92 | |
GenBank mRNA | BC058154.1 | - | 3 - 4 | 357 | 92 | |
BC088312.1 | - | 3 - 4 | 353 | 7324682 | 92 | |
FQ212609.1 | - | 3 - 4 | 363 | 92 | ||
FQ214728.1 | - | 1 - 2 | 141 | 92 | ||
FQ215871.1 | - | 3 - 4 | 365 | 92 | ||
FQ219747.1 | - | 3 - 4 | 351 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |