Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108505.2 | NP_001101975.2 | 6 - 7 | 798 | 67 | |
GenBank mRNA | FQ228402.1 | - | 6 - 7 | 796 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |