Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033757.1 | NP_001028929.1 | 2 - 3 | 1346 | 58 | |
NM_001033758.1 | NP_001028930.1 | 2 - 3 | 1310 | 58 | ||
NM_182735.2 | NP_877399.2 | 3 - 4 | 1175 | 58 | ||
XM_008760076.1 | XP_008758298.1 | 3 - 4 | 1258 | 58 | ||
XM_008760078.1 | XP_008758300.1 | 3 - 4 | 1218 | 58 | ||
XM_008760079.1 | XP_008758301.1 | 3 - 4 | 1216 | 58 | ||
GenBank mRNA | AY351678.1 | - | 2 - 3 | 1347 | 58 | |
AY351679.1 | - | 2 - 3 | 1311 | 58 | ||
AY576608.1 | - | 3 - 4 | 1174 | 58 | ||
AY586728.1 | - | 3 - 4 | 1139 | 58 | ||
BC098646.1 | - | 3 - 4 | 1173 | 7109418 | 58 | |
CF110214.1 | - | 2 - 3 | 491 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |