Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053610.1 | NP_446062.1 | 2 - 3 | 345 | 57 | |
GenBank mRNA | AF110732.1 | - | 2 - 3 | 345 | 57 | |
BC078771.1 | - | 2 - 3 | 321 | 7193191 | 57 | |
FQ210686.1 | - | 2 - 3 | 276 | 57 | ||
FQ223238.1 | - | 2 - 3 | 273 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |