Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760198.1 | XP_008758420.1 | 5 - 6 | 1576 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |