Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127543.1 | NP_001121015.1 | 3 - 4 | 508 | 67 | |
XM_006230811.2 | XP_006230873.1 | 3 - 4 | 999 | 67 | ||
GenBank mRNA | BC166607.1 | - | 3 - 4 | 508 | 5598287 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |