Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prune2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231152.2 | XP_006231214.2 | 15 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |