Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138908.2 | NP_620263.1 | 4 - 5 | 914 | 102 | |
GenBank mRNA | Y09945.1 | - | 4 - 5 | 916 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |