Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031153.2 | NP_112415.1 | 7 - 8 | 1376 | 87 | |
GenBank mRNA | AF203906.1 | - | 7 - 8 | 1050 | 87 | |
BC083553.1 | - | 7 - 8 | 1376 | 7189111 | 87 | |
FQ213170.1 | - | 7 - 8 | 994 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |