Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022860.2 | NP_074051.1 | 5 - 6 | 672 | 67 | |
XM_006241497.2 | XP_006241559.1 | 5 - 6 | 1199 | 67 | ||
XM_008765419.1 | XP_008763641.1 | 5 - 6 | 3028 | 67 | ||
XM_008765420.1 | XP_008763642.1 | 4 - 5 | 1349 | 67 | ||
GenBank mRNA | BC081799.1 | - | 5 - 6 | 672 | 7133837 | 67 |
D17809.1 | - | 5 - 6 | 561 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |