Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012887.1 | NP_037019.1 | 3 - 4 | 808 | 100 | |
XM_008765219.1 | XP_008763441.1 | 3 - 4 | 847 | 100 | ||
XM_008765220.1 | XP_008763442.1 | 3 - 4 | 847 | 100 | ||
GenBank mRNA | U18314.1 | - | 3 - 4 | 808 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |