Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053311.1 | NP_445763.1 | 20 - 21 | 3607 | 98 | |
XM_006241294.2 | XP_006241356.1 | 20 - 21 | 3699 | 98 | ||
XM_006241295.2 | XP_006241357.1 | 19 - 20 | 3576 | 98 | ||
GenBank mRNA | J03753.1 | - | 20 - 21 | 3607 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |