Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199098.1 | NP_954529.1 | 1 - 2 | 419 | 91 | |
XM_006241347.2 | XP_006241409.1 | 2 - 3 | 516 | 91 | ||
XM_006241348.2 | XP_006241410.1 | 2 - 3 | 369 | 91 | ||
XM_006241349.1 | XP_006241411.1 | 2 - 3 | 254 | 91 | ||
XM_008765355.1 | XP_008763577.1 | 1 - 2 | 465 | 91 | ||
GenBank mRNA | BC061871.1 | - | 1 - 2 | 419 | 5598899 | 91 |
Gene Symbol: LOC100912615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765371.1 | XP_008763593.1 | 524 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |