Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106775.1 | NP_001100245.1 | 7 - 8 | 960 | 143 | |
XM_006241197.1 | XP_006241259.1 | 7 - 8 | 865 | 143 | ||
XM_006241198.2 | XP_006241260.1 | 7 - 8 | 860 | 143 | ||
GenBank mRNA | BC091253.1 | - | 6 - 7 | 722 | 143 | |
BC161937.1 | - | 7 - 8 | 832 | 9022830 | 143 | |
FQ220712.1 | - | 7 - 8 | 843 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |